Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gpd1
Entrez Gene ID
448519
Gene Name
glycerol-3-phosphate dehydrogenase 1
Gene Aliases
-
Chromosome Location
Chr.2: 145950330 - 145968336 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006807.1 NP_001006808.1 6 - 7 893 69
GenBank mRNA BC076683.1 - 6 - 7 893 69
CR761150.2 - 6 - 7 915 69

View Full Product Details