Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
smarcb1
Entrez Gene ID
448543
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1
Gene Aliases
baf47, hsnfs, ini1, rdt, sfh1p, snf5, snf5l1, snr1
Chromosome Location
Chr.1: 144660459 - 144677925 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006818.1 NP_001006819.1 3 - 4 341 66
XM_012959983.2 XP_012815437.1 3 - 4 380 66
GenBank mRNA BC076714.1 - 3 - 4 341 66

View Full Product Details