Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crabp2
Entrez Gene ID
448629
Gene Name
cellular retinoic acid binding protein 2
Gene Aliases
crabp-ii, rbp6, xCRABP
Chromosome Location
Chr.8: 111001960 - 111023441 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006862.1 NP_001006863.1 2 - 3 402 92
GenBank mRNA BC076971.1 - 2 - 3 402 92

View Full Product Details