Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bcl7a
Entrez Gene ID
448642
Gene Name
B-cell CLL/lymphoma 7A
Gene Aliases
-
Chromosome Location
Chr.1: 153411449 - 153426307 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006871.1 NP_001006872.1 2 - 3 244 77
GenBank mRNA BC076987.1 - 2 - 3 244 77
CR926398.1 - 2 - 3 185 77

View Full Product Details