Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eef1b2
Entrez Gene ID
448658
Gene Name
eukaryotic translation elongation factor 1 beta 2
Gene Aliases
eef1b, ef1b
Chromosome Location
Chr.9: 48458589 - 48462472 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006876.2 NP_001006877.1 1 - 2 116 89
GenBank mRNA BC077005.1 - 1 - 2 117 89

View Full Product Details