Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
haao
Entrez Gene ID
448761
Gene Name
3-hydroxyanthranilate 3,4-dioxygenase
Gene Aliases
-
Chromosome Location
Chr.5: 13367580 - 13437843 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006913.1 NP_001006914.1 5 - 6 526 108
XM_018093843.1 XP_017949332.1 6 - 7 469 108
XM_018093844.1 XP_017949333.1 6 - 7 462 108
GenBank mRNA BC075395.1 - 5 - 6 526 108
BC087758.1 - 5 - 6 451 108

View Full Product Details