Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmem248
Entrez Gene ID
493220
Gene Name
transmembrane protein 248
Gene Aliases
c7orf42
Chromosome Location
Chr.2: 44353376 - 44363646 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007493.1 NP_001007494.1 2 - 3 588 101
GenBank mRNA BC079930.1 - 2 - 3 588 101
CR762290.2 - 2 - 3 545 101

View Full Product Details