Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tmem248
Entrez Gene ID
493220
Gene Name
transmembrane protein 248
Gene Aliases
c7orf42
Chromosome Location
Chr.2: 44353376 - 44363646 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007493.1 NP_001007494.1 5 - 6 1065 106
GenBank mRNA BC079930.1 - 5 - 6 1065 106
CR762290.2 - 5 - 6 1022 106

View Full Product Details