Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c20orf27
Entrez Gene ID
493230
Gene Name
chromosome 20 open reading frame 27
Gene Aliases
-
Chromosome Location
Chr.1: 9459311 - 9486910 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007503.1 NP_001007504.1 1 - 2 292 86
GenBank mRNA BC079940.1 - 1 - 2 292 86

View Full Product Details