Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps2
Entrez Gene ID
493255
Gene Name
ribosomal protein S2
Gene Aliases
-
Chromosome Location
Chr.9: 11142149 - 11146148 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007868.1 NP_001007869.1 3 - 4 347 83
XM_018097089.1 XP_017952578.1 3 - 4 369 83
GenBank mRNA BC061291.1 - 3 - 4 347 83
BC080133.1 - 3 - 4 347 83
CR855816.2 - 3 - 4 345 83

View Full Product Details