Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cbr4
Entrez Gene ID
493259
Gene Name
carbonyl reductase 4
Gene Aliases
-
Chromosome Location
Chr.1: 40968815 - 40977977 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007872.1 NP_001007873.1 5 - 6 567 98
XM_012960249.2 XP_012815703.1 5 - 6 645 98
XM_012960250.2 XP_012815704.1 5 - 6 794 98
XM_012960257.2 XP_012815711.1 5 - 6 730 98
GenBank mRNA BC080139.1 - 5 - 6 567 98

View Full Product Details