Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arl15
Entrez Gene ID
493266
Gene Name
ADP ribosylation factor like GTPase 15
Gene Aliases
arfrp2
Chromosome Location
Chr.1: 179157393 - 179335147 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007880.1 NP_001007881.1 4 - 5 596 78
XM_012960270.2 XP_012815724.1 5 - 6 901 78
XM_012960277.2 XP_012815731.1 4 - 5 844 78
GenBank mRNA BC080149.1 - 4 - 5 596 78

View Full Product Details