Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab25
Entrez Gene ID
493286
Gene Name
RAB25, member RAS oncogene family
Gene Aliases
catx-8
Chromosome Location
Chr.8: 111854301 - 111869826 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007902.1 NP_001007903.1 4 - 5 700 67
GenBank mRNA BC080339.1 - 4 - 5 700 67
CR848299.2 - 4 - 5 689 67

View Full Product Details