Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pcna
Entrez Gene ID
493302
Gene Name
proliferating cell nuclear antigen
Gene Aliases
-
Chromosome Location
Chr.3: 111204678 - 111208820 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007920.1 NP_001007921.1 4 - 5 695 92
GenBank mRNA BC080365.1 - 4 - 5 695 92

View Full Product Details