Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
adipor1
Entrez Gene ID
493308
Gene Name
adiponectin receptor 1
Gene Aliases
acdcr1, cgi-45, cgi45, paqr1, tesbp1a
Chromosome Location
Chr.2: 49304836 - 49318142 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007927.1 NP_001007928.1 6 - 7 1008 88
XM_012960344.2 XP_012815798.1 6 - 7 1003 88
XM_012960345.2 XP_012815799.1 6 - 7 1012 88
GenBank mRNA BC080374.1 - 6 - 7 1008 88

View Full Product Details