Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpsf4
Entrez Gene ID
493312
Gene Name
cleavage and polyadenylation specific factor 4
Gene Aliases
cpsf30, nar, neb1
Chromosome Location
Chr.9: 38906468 - 38916821 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007932.1 NP_001007933.1 7 - 8 901 65
XM_012970321.2 XP_012825775.1 6 - 7 674 65
XM_012970322.2 XP_012825776.1 6 - 7 709 65
GenBank mRNA BC080440.1 - 7 - 8 901 65
CR761660.2 - 6 - 7 615 65

View Full Product Details