Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
lin37
Entrez Gene ID
493362
Gene Name
lin-37 DREAM MuvB core complex component
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683384: 955631 - 964625 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007999.1 NP_001008000.1 7 - 8 857 83
GenBank mRNA BC080878.1 - 7 - 8 857 83

View Full Product Details