Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
adprm
Entrez Gene ID
493375
Gene Name
ADP-ribose/CDP-alcohol diphosphatase, manganese-dependent
Gene Aliases
c17orf48
Chromosome Location
Chr.10: 38309916 - 38316458 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008012.1 NP_001008013.1 2 - 3 663 75
XM_012971669.2 XP_012827123.1 2 - 3 729 75
GenBank mRNA BC080891.1 - 2 - 3 663 75

View Full Product Details