Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spred1
Entrez Gene ID
493399
Gene Name
sprouty related, EVH1 domain containing 1
Gene Aliases
Xspred1, Xtspred1
Chromosome Location
Chr.8: 90501109 - 90551511 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008036.1 NP_001008037.1 5 - 6 708 65
GenBank mRNA AY714338.1 - 5 - 6 708 65
BC080919.1 - 5 - 6 708 65

View Full Product Details