Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tada3
Entrez Gene ID
493402
Gene Name
transcriptional adaptor 3
Gene Aliases
ada3, ngg1, tada3-a, tada3-b, tada3l
Chromosome Location
Chr.4: 128008916 - 128018679 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008039.1 NP_001008040.1 3 - 4 557 72
XM_012960931.1 XP_012816385.1 3 - 4 462 72
XM_018093018.1 XP_017948507.1 3 - 4 431 72
GenBank mRNA BC080923.1 - 3 - 4 557 72

View Full Product Details