Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ears2
Entrez Gene ID
493404
Gene Name
glutamyl-tRNA synthetase 2, mitochondrial
Gene Aliases
mse1
Chromosome Location
Chr.9: 16755004 - 16760226 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008041.1 NP_001008042.1 8 - 9 1497 57
GenBank mRNA BC080925.1 - 8 - 9 1497 57

View Full Product Details