Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
churc1
Entrez Gene ID
493426
Gene Name
churchill domain containing 1
Gene Aliases
chch, churchill, my015
Chromosome Location
Chr.8: 87422727 - 87429934 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008063.1 NP_001008064.1 1 - 2 124 89
GenBank mRNA BC080951.1 - 1 - 2 124 89

View Full Product Details