The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008083.1 | NP_001008084.1 | 6 - 7 | 951 | 85 |
GenBank mRNA | BC080983.1 | - | 6 - 7 | 951 | 85 |
CR926407.2 | - | 6 - 7 | 869 | 85 |