Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apoa4
Entrez Gene ID
493461
Gene Name
apolipoprotein A4
Gene Aliases
myh11
Chromosome Location
Chr.7: 83943422 - 83947618 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008098.2 NP_001008099.1 1 - 2 75 65
XM_012966475.1 XP_012821929.1 2 - 3 546 65
GenBank mRNA BC081300.1 - 1 - 2 79 65

View Full Product Details