Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
grb2
Entrez Gene ID
493492
Gene Name
growth factor receptor bound protein 2
Gene Aliases
EGFRBP-GRB2, Grb3-3, MST084, MSTP084, grb2-a, grb2-b
Chromosome Location
Chr.10: 35858612 - 35885269 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008129.1 NP_001008130.1 3 - 4 349 104
XM_012971685.2 XP_012827139.1 3 - 4 437 104
XM_012971686.2 XP_012827140.1 3 - 4 418 104
XM_012971687.2 XP_012827141.1 3 - 4 353 104
XM_012971688.2 XP_012827142.1 3 - 4 353 104
XM_012971689.2 XP_012827143.1 3 - 4 437 104
GenBank mRNA BC081338.1 - 3 - 4 349 104

View Full Product Details