Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kpna2
Entrez Gene ID
493517
Gene Name
karyopherin alpha-2 subunit like
Gene Aliases
ipoa1, qip2, rch1, srp1alpha
Chromosome Location
Chr.Un|NW_016683548: 178908 - 185103 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008154.2 NP_001008155.1 2 - 3 138 77
XM_012966479.2 XP_012821933.1 2 - 3 231 77
GenBank mRNA BC081368.1 - 2 - 3 140 77
CR762026.2 - 1 - 2 111 77

View Full Product Details