Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
crisp2
Entrez Gene ID
493566
Gene Name
cysteine-rich secretory protein 2
Gene Aliases
crisp-2, ct36, gapdl5, tpx1, tsp1
Chromosome Location
Chr.5: 144862451 - 144875933 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008203.1 NP_001008204.1 5 - 6 450 115
GenBank mRNA BC080935.1 - 5 - 6 450 115
CT025286.2 - 5 - 6 439 115

View Full Product Details