Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acp5
Entrez Gene ID
493572
Gene Name
acid phosphatase 5, tartrate resistant
Gene Aliases
TRAP-5b
Chromosome Location
Chr.3: 128526876 - 128547543 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008209.1 NP_001008210.1 2 - 3 328 108
XM_012953156.2 XP_012808610.1 2 - 3 304 108
GenBank mRNA BC081357.1 - 2 - 3 328 108
CR942420.2 - 2 - 3 301 108

View Full Product Details