Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lrrc20
Entrez Gene ID
496473
Gene Name
leucine rich repeat containing 20
Gene Aliases
-
Chromosome Location
Chr.7: 32606523 - 32724817 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011063.1 NP_001011063.1 4 - 5 457 81
XM_012966493.2 XP_012821947.1 5 - 6 797 81
GenBank mRNA BC084442.1 - 4 - 5 457 81

View Full Product Details