Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c5orf22
Entrez Gene ID
496505
Gene Name
chromosome 5 open reading frame 22
Gene Aliases
-
Chromosome Location
Chr.6: 54077418 - 54139817 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011092.1 NP_001011092.1 5 - 6 867 181
GenBank mRNA BC084484.1 - 5 - 6 867 181

View Full Product Details