Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hnrnpa1
Entrez Gene ID
496507
Gene Name
heterogeneous nuclear ribonucleoprotein A1
Gene Aliases
hnrpa1
Chromosome Location
Chr.2: 142403036 - 142411342 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011094.1 NP_001011094.1 4 - 5 532 87
XM_012956844.1 XP_012812298.1 4 - 5 686 87
XM_012956845.2 XP_012812299.1 4 - 5 567 87
GenBank mRNA BC084487.1 - 4 - 5 532 87
CT010562.2 - 4 - 5 495 87

View Full Product Details