Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
snx10
Entrez Gene ID
496527
Gene Name
sorting nexin 10
Gene Aliases
-
Chromosome Location
Chr.6: 33392962 - 33414564 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011114.2 NP_001011114.1 4 - 5 294 96
XM_012964580.2 XP_012820034.1 4 - 5 374 96
XM_012964581.2 XP_012820035.1 4 - 5 386 96
XM_012964582.2 XP_012820036.1 3 - 4 377 96
GenBank mRNA BC084512.1 - 4 - 5 295 96
CR760982.2 - 4 - 5 265 96
CR761975.1 - 4 - 5 287 96

View Full Product Details