Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
snx10
Entrez Gene ID
496527
Gene Name
sorting nexin 10
Gene Aliases
-
Chromosome Location
Chr.6: 33392962 - 33414564 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011114.2 NP_001011114.1 5 - 6 402 68
XM_012964580.2 XP_012820034.1 5 - 6 482 68
XM_012964581.2 XP_012820035.1 5 - 6 494 68
XM_012964582.2 XP_012820036.1 4 - 5 485 68
GenBank mRNA BC084512.1 - 5 - 6 403 68
CR760982.2 - 5 - 6 373 68
CR761975.1 - 5 - 6 395 68

View Full Product Details