Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pcdh18
Entrez Gene ID
496568
Gene Name
protocadherin 18
Gene Aliases
pcdh68l
Chromosome Location
Chr.1: 52730305 - 52739775 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011150.2 NP_001011150.1 1 - 2 2941 78
GenBank mRNA BC084899.1 - 1 - 2 2942 78

View Full Product Details