Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cplx3
Entrez Gene ID
496634
Gene Name
complexin 3
Gene Aliases
cpx-iii, cpxiii, lman1
Chromosome Location
Chr.3: 84361604 - 84391446 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011205.1 NP_001011205.1 3 - 4 463 97
GenBank mRNA BC087752.1 - 3 - 4 463 97

View Full Product Details