Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bhmt
Entrez Gene ID
496651
Gene Name
betaine--homocysteine S-methyltransferase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016684775: 119 - 9739 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011217.1 NP_001011217.1 2 - 3 170 72
GenBank mRNA BC087774.1 - 2 - 3 170 72
BC089235.1 - 2 - 3 154 72

View Full Product Details