Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
esd
Entrez Gene ID
496692
Gene Name
esterase D
Gene Aliases
-
Chromosome Location
Chr.2: 131274374 - 131307081 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011247.1 NP_001011247.1 2 - 3 143 92
XM_018091202.1 XP_017946691.1 2 - 3 888 92
GenBank mRNA BC087823.1 - 2 - 3 143 92
CT030509.1 - 2 - 3 106 92

View Full Product Details