Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acta2
Entrez Gene ID
496696
Gene Name
actin, alpha 2, smooth muscle, aorta
Gene Aliases
ASMA, SMactin, a-sma, aat6, actc1, actsa, alpha-actin, mymy5, sm-actin, sma
Chromosome Location
Chr.7: 4480359 - 4487475 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011250.1 NP_001011250.1 5 - 6 665 58
GenBank mRNA BC087829.1 - 5 - 6 665 58
CU025168.1 - 6 - 7 675 58

View Full Product Details