Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
psma3
Entrez Gene ID
496707
Gene Name
proteasome subunit alpha 3
Gene Aliases
-
Chromosome Location
Chr.8: 83300783 - 83315288 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011257.1 NP_001011257.1 8 - 9 651 91
XM_012968299.2 XP_012823753.1 7 - 8 673 91
GenBank mRNA BC087567.2 - 8 - 9 651 91
CR855724.2 - 8 - 9 622 91

View Full Product Details