Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
serpinb5
Entrez Gene ID
496735
Gene Name
serpin peptidase inhibitor, clade B (ovalbumin), member 5
Gene Aliases
-
Chromosome Location
Chr.6: 52434714 - 52459114 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011282.1 NP_001011282.1 5 - 6 633 119
GenBank mRNA BC087996.1 - 5 - 6 633 119

View Full Product Details