Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rac2
Entrez Gene ID
496738
Gene Name
Rac family small GTPase 2
Gene Aliases
en-7, hspc022
Chromosome Location
Chr.4: 112651699 - 112677222 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011285.1 NP_001011285.1 2 - 3 154 106
XM_012961000.2 XP_012816454.1 2 - 3 229 106
GenBank mRNA BC087999.1 - 2 - 3 154 106
CR760806.2 - 2 - 3 138 106

View Full Product Details