Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zdhhc3
Entrez Gene ID
496743
Gene Name
zinc finger DHHC-type containing 3
Gene Aliases
godz, znf373
Chromosome Location
Chr.6: 63691682 - 63720966 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011290.1 NP_001011290.1 2 - 3 469 94
XM_012964606.2 XP_012820060.1 2 - 3 424 94
XM_012964607.2 XP_012820061.1 2 - 3 531 94
XM_012964608.2 XP_012820062.1 2 - 3 605 94
XM_012964609.2 XP_012820063.1 2 - 3 519 94
XM_012964610.2 XP_012820064.1 2 - 3 605 94
GenBank mRNA BC088007.1 - 2 - 3 469 94
CR848357.2 - 2 - 3 408 94

View Full Product Details