Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rps4x
Entrez Gene ID
394507
Gene Name
ribosomal protein S4 X-linked
Gene Aliases
ccg2, dxs306, rps4, scar, scr10
Chromosome Location
Chr.8: 18449861 - 18469617 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203581.3 NP_988912.1 5 - 6 828 68
XM_012968148.1 XP_012823602.1 6 - 7 750 68
GenBank mRNA BC059771.1 - 4 - 5 536 68
BC077671.1 - 5 - 6 831 68
CR760832.2 - 4 - 5 536 68

View Full Product Details