Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
snf8
Entrez Gene ID
496607
Gene Name
SNF8, ESCRT-II complex subunit
Gene Aliases
d11moh34, vps22
Chromosome Location
Chr.10: 5258761 - 5264457 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011185.2 NP_001011185.1 6 - 7 583 107
GenBank mRNA BC086504.2 - 6 - 7 583 107

View Full Product Details