Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
scn4b
Entrez Gene ID
496766
Gene Name
sodium voltage-gated channel beta subunit 4
Gene Aliases
-
Chromosome Location
Chr.7: 84739740 - 84767197 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011311.1 NP_001011311.1 3 - 4 374 76
GenBank mRNA BC088037.1 - 3 - 4 374 76

View Full Product Details