Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atraid
Entrez Gene ID
496775
Gene Name
all-trans retinoic acid induced differentiation factor
Gene Aliases
apr-3, apr3, c2orf28
Chromosome Location
Chr.Un|NW_016687619: 585 - 3074 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011316.1 NP_001011316.1 - 354 99
GenBank mRNA BC088051.1 - - 354 99
CU075626.1 - 2 - 3 350 99

View Full Product Details