Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gipc1
Entrez Gene ID
496793
Gene Name
GIPC PDZ domain containing family member 1
Gene Aliases
GIPC, Kermit, XGIPC, gipc3, glut1cbp, iip-1, rgs19ip1, semcap, synectiin
Chromosome Location
Chr.3: 124305731 - 124315552 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011331.1 NP_001011331.1 4 - 5 799 79
XM_012953129.2 XP_012808583.1 4 - 5 932 79
GenBank mRNA BC088477.1 - 4 - 5 799 79
CR760513.2 - 4 - 5 802 79

View Full Product Details