Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vapa
Entrez Gene ID
496796
Gene Name
VAMP associated protein A
Gene Aliases
vap-a, vap33
Chromosome Location
Chr.6: 89550546 - 89570355 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011333.1 NP_001011333.1 4 - 5 692 101
GenBank mRNA BC088480.1 - 4 - 5 692 101
CR761106.2 - 4 - 5 665 101

View Full Product Details