Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrpl44
Entrez Gene ID
496841
Gene Name
mitochondrial ribosomal protein L44
Gene Aliases
-
Chromosome Location
Chr.5: 125194838 - 125201299 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011374.1 NP_001011374.1 1 - 2 165 82
GenBank mRNA BC088549.1 - 1 - 2 165 82

View Full Product Details