Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zmynd12
Entrez Gene ID
496847
Gene Name
zinc finger MYND-type containing 12
Gene Aliases
-
Chromosome Location
Chr.7: 77955149 - 77967549 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011379.1 NP_001011379.1 2 - 3 309 92
GenBank mRNA BC088560.1 - 2 - 3 309 92
CR848399.2 - 2 - 3 312 92

View Full Product Details